missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEX26 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27472-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene belongs to the peroxin-26 gene family. It is probably required for protein import into peroxisomes. It anchors PEX1 and PEX6 to peroxisome membranes, possibly to form heteromeric AAA ATPase complexes required for the import of proteins into peroxisomes. Defects in this gene are the cause of peroxisome biogenesis disorder complementation group 8. PBD refers to a group of peroxisomal disorders arising from a failure of protein import into the peroxisomal membrane or matrix. The PBD group is comprised of four disorders: Zellweger syndrome, neonatal adrenoleukodystrophy, infantile Refsum disease, and classical rhizomelic chondrodysplasia punctata. Alternatively spliced transcript variants have been identified for this gene.Specifications
| PEX26 | |
| Polyclonal | |
| Unconjugated | |
| PEX26 | |
| Peroxin 26, Peroxisome assembly protein 26, PEX26, PEX26M1T, Pex26pM1T | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55670 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q7Z412 | |
| PEX26 | |
| PEX26 Fusion Protein Ag26529 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction