missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEX12 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27011-1-AP-150UL
This item is not returnable.
View return policy
Description
Peroxins (PEXs) are proteins that are essential for the assembly of functional peroxisomes. The peroxisome biogenesis disorders (PBDs) are a group of genetically heterogeneous autosomal recessive, lethal diseases characterized by multiple defects in peroxisome function. The peroxisomal biogenesis disorders are a heterogeneous group with at least 14 complementation groups and with more than 1 phenotype being observed in cases falling into particular complementation groups. Although the clinical features of PBD patients vary, cells from all PBD patients exhibit a defect in the import of one or more classes of peroxisomal matrix proteins into the organelle.Specifications
| PEX12 | |
| Polyclonal | |
| Unconjugated | |
| PEX12 | |
| PAF 3, PAF3, Peroxin 12, Peroxisome assembly factor 3, Peroxisome assembly protein 12, PEX12 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5193 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O00623 | |
| PEX12 | |
| PEX12 Fusion Protein Ag25500 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction