Learn More
PEX1 Mouse anti-Human, Unconjugated, Clone: 2F3H5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67436-1-IG-20UL
 View more versions of this product
Product Code. 16860066
  • 1710.33 NOK / 20µL
Description
The PEX1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PEX1. This antibody recognizes human, mouse, rat antigen. The PEX1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
PEX1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, ELISA
2F3H5
Unconjugated
20 μL
5189
IgG2b