Learn More
Corning™ PETG Baffled Erlenmeyer Flask with Vent Cap
BIOPHARMAGREENER_CHOICE
Product Code. 15863875 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
2 L
3 L
5 L
Unit Size:
Pack of 4
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15863875 2 L Pack of 6
15883875 3 L Pack of 4
15803885 5 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15863875 Supplier Corning™ Supplier No. 431281
  • ONLINE EXCLUSIVE PRICE
    3555.00 NOK
    3318.16 NOK / Pack of 6
    Save 236.84 NOK
    6% Off
Estimated Shipment: 05-01-2027
Add to basket
Add to basket
Constructed of optically clear polyethylene terephthalate (PETG) and ideal for shaker culture applications.
The Corning™ Erlenmeyer flask is designed to help scale-up suspension culture with space efficiency in mind. The flasks are ready-to-use, ideal for shaker culture applications, and storage.
  • Available with baffled bottom
  • Molded-in graduations for accuracy
  • Large vent cap for continuous gas exchange while preventing leakage
  • Highest Sterility Assurance Level (SAL) of 10−6
  • Nonpyrogenic and RNase-/DNase-free
  • Made from optically-clear polyethylene terephthalate(PTEG)
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Nonpyrogenic, DNase/RNase-free
Closure Color Orange
Closure Material Polypropylene
Capacity (Metric) 2 L
Material PETG
For Use With (Application) Suspension Cell Cultures
For Use With (Equipment) Orbital Shaker
No. per Case 6/Cs.
No. per Pack 1/Pk.
Product Type Cell Culture Erlenmeyer Flask
Shape Baffled Erlenmeyer
Disposable Single-use
Graduated Yes
Mouth Style Threaded
Sterility Sterile
Bottom Shape Baffled
Closure Type Vented
Show More Show Less
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.