Learn More
Haldenwanger™ Pestle
Product Code. 12021812
Change view
Click to view available options
Diameter (Metric) Outer:
24mm
28mm
30mm
42mm
45mm
48mm
Unit Size:
Each
Pack of 10
Pack of 5
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
12021812 30mm Each
10597644 48mm Pack of 5
10480724 28mm Pack of 10
10245164 45mm Pack of 5
12081802 24mm Each
10744585 42mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Product Code. 12021812 Supplier Haldenwanger™ Supplier No. L 56/1/GLASIERT
 Restricted Item
Estimated Shipment: 19-10-2026
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Pestle
Grinding surfaces are unglazed.
Specifications
Diameter (Metric) Outer 30mm
Product Type Pestle
Length (Metric) 135mm
Type Pestle