Learn More
Pesticides Surrogate Standard Spiking Solution 200 μg/mL in Acetone, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 2,4,5,6-Tetrachloro-m-xylene∣PCB No. 209 |
| CAS | 877-09-8, 2051-24-3 |
| Analyte or Component Names | 2,4,5,6-Tetrachloro-m-xylene∣PCB No. 209 |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetone |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 2,4,5,6-Tetrachloro-m-xylene 200 μg/mL∣PCB No. 209 200 μg/mL |