Learn More
Pesticide Mix 7, SPEX CertiPrep™
Product Code. 16340488 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
16340488 1 mL 1mL
1 options
Product Code. 16340488
Brand: SPEX Certiprep SPXPR7
Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Mix 7 containing 8 compounds in acetronitrile is ideal for pesticide screening, food safety and QuEChERS applications.
  • Pesticide Mix 7 containing 8 compounds in acetronitrile
  • Available and sold in 1mL ampules that are supplied with a pre-labeled transfer/storage vial
Warranty
1 year
Specifications
Chemical Name or Material Cypermethrin and an isomer zeta-cypermethrin,
Cyfluthrin (baythroid)
Bifenthrin
Tetramethri
Packaging Glass Ampule
CAS 52645-53-1, 8003-34-7, 10453-86-8, 7696-12-0, 23031-36-9
Assay Percent Range 98.4
99
98
98.2
97.7
98.1
99.4
25
Molecular Formula C22H19Cl2NO3
C22H18Cl2FNO3
C23H22ClF3O2
C19H25NO4
C19H24O3
C21H20Cl2O3
C22H26O3
C22H28O5
Concentration 100 μg/mL
Quantity 1 mL
Formula Weight 416.297 g/mol
434.2876 g/mol
422.8678 g/mol
331.4061 g/mol
300.392 g/mol
391.288 g/mol
338.44 g/mol
372.4547 g/mol
compliance-icons compliance-icons
Product Identifier
  • Pesticide Mix 7
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : acetonitrile