missing translation for 'onlineSavingsMsg'
Learn More

Peroxyacetic Acid, 3.5% (w/w), Ricca

Product Code. 31940687
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31940687 500 mL 500mL
31940561 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 31940687 Supplier Ricca Chemical Company Supplier No. R5495000500C
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
  • Peroxyacetic Acid
  • Available in 500 mL and 1 L package sizes in amber glass containers

Chemical Identifiers

Packaging Amber Glass Container

Specifications

Color-coded No
Form Liquid
Scent Unscented
Sterility Non-sterile
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.