Learn More
Thermo Scientific Chemicals Peroxidase, horseradish
Product Code. 15402879
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
Product Code. Quantity unitSize
15402879 100 mg 100mg
15492869 1 g 1g
2 options
Product Code. 15402879
Brand: Thermo Scientific Chemicals J60026.MC
Request Bulk or Custom Format
Used in the oxidation of luminol to 3-aminophthalate. It is often used in conjugates to determine the presence of a molecular target, especially in the detection of specific protein in a western blot after conjugation with antibody.
Peroxidase, horseradish is used in the oxidation of luminol to 3-aminophthalate. It is often used in conjugates to determine the presence of a molecular target, especially in the detection of specific protein in a western blot after conjugation with antibody. Further, it is highly reactive toward human tumor cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 mg
Form Lyophilized
What is the solubility of Peroxidase, horseradish (Cat. No. J60026.03, J60026.MC)?
Peroxidase, horseradish is soluble in water or 100mM phosphate buffer.