Learn More
Brady™ Permanent Self-laminating Vinyl Wrap Around Wire and Cable Labels for M6/M7 Printers
Product Code. 17861609 Shop All Brady Products
Change view
Click to view available options
Dimensions (L x W):
0.25 x 1.1 in.
1 x 1.9 in.
Height (Metric):
6.35 mm
25.4 mm
Unit Size:
Pack of 250
Pack of 750
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Height (Metric) Unit Size
17861609 1 x 1.9 in. 25.4 mm Pack of 250
17831649 0.25 x 1.1 in. 6.35 mm Pack of 750
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17861609 Supplier Brady™ Supplier No. 173903
Support a Safer, More Efficient Laboratory
Use promo code 28026
View all promotions
Estimated Shipment: 01-12-2026
Self-laminating Vinyl is an ideal material for wire and cable identification. This material has good clarity, conformability and is self-extinguishing.
  • Self-laminating vinyl is ideal for use in cable and wire labeling and general-purpose marking in labs
  • Durable, permanent acrylic adhesive has outstanding water and oil resistance
  • UL approved
  • Service temperature of -40°C to 70°C (-40°F to 158°F)
  • Made of B-427 vinyl
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized
Adhesive Type Permanent Acrylic
Coating None
Color White
Finish Matte
Format Die-cut
Material Vinyl
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Data and Telecommunications Labeling, Electrical Labeling, Laboratory Labeling, Wire and Cable Labeling
For Use With (Equipment) M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
Latex Free No
Quantity 250 Labels
Shape Rectangle
Temperature Range (English) -94°F to +158°F
Temperature Range (Metric) -70°C to +70°C
Thickness (English) 0.004 in.
Thickness (Metric) 0.102 mm
Dimensions (L x W) 1 x 1.9 in.
Length (English) 1 in.
Width (English) 1.9 in.
Width (Metric) 48.26 mm
Height (Metric) 25.4 mm
Alternate Part Number M6-78-427-AW