Learn More
Brady™ Permanent Paint
Description
- Floor marking paint.
Specifications
Specifications
| Quantity | 6 Pk. |
| Alternate Part Number | EASYLINE PAINT BLACK (PAC of 6) |
| Product Type | Paint |
| For Use With (Application) | Floor marking |
| Color | Black |