Learn More
Macherey-Nagel™ Permabond™ Silane GC Capillary Column
Product Code. 10651492 Shop All Macherey-Nagel Products
Click to view available options
Length (Metric):
25 m
50 m
Unit Size:
Each
Product Code. 10651492
Brand: Macherey-Nagel™ 723411.25
Only null left
Specifications
Diameter (Metric) 0.8 mm
Type GC Capillary Column
Diameter (Metric) Inner 0.53 mm
Stationary Phase Permabond™ Silane
For Use With (Application) Monomeric Silanes and Chlorosilanes Analysis
Temperature Range 260°C to 280°C
Length (Metric) 25 m
Packing Material Permabond™ Silane
Quantity 1 Each