Learn More
Macherey-Nagel™ Permabond™ FFAP GC Capillary Column
Product Code. 10727472 Shop All Macherey-Nagel Products
Change view
Click to view available options
Length (Metric):
10 m
20 m
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.1 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.1 μm
0.25 μm
0.35 μm
0.5 μm
1 μm
Unit Size:
Each
Pack of 10
Product Code. Length (Metric) Diameter (Metric) Inner Film Thickness unitSize
10727472 25 m 0.53 mm 1 μm Each
10786122 10 m 0.1 mm 0.1 μm Each
10601112 10 m 0.25 mm 0.25 μm Pack of 10
10586501 10 m 0.1 mm 0.25 μm Each
10033011 10 m 0.32 mm 0.35 μm Each
10348641 10 m 0.32 mm 0.5 μm Each
10398301 10 m 0.53 mm 1 μm Each
10236801 20 m 0.1 mm 0.1 μm Each
10698801 25 m 0.25 mm 0.1 μm Each
10368541 25 m 0.25 mm 0.25 μm Each
10440512 25 m 0.32 mm 0.1 μm Each
10556341 25 m 0.32 mm 0.25 μm Each
10318641 25 m 0.32 mm 0.35 μm Each
10576861 25 m 0.32 mm 0.5 μm Each
10479931 30 m 0.25 mm 0.25 μm Each
10469741 30 m 0.32 mm 0.25 μm Each
10338021 30 m 0.32 mm 0.5 μm Each
10023011 50 m 0.25 mm 0.25 μm Each
10796122 50 m 0.25 mm 0.1 μm Each
10708242 50 m 0.32 mm 0.25 μm Each
10092851 50 m 0.32 mm 0.35 μm Each
10348211 50 m 0.32 mm 0.1 μm Each
10065841 50 m 0.32 mm 0.5 μm Each
10072961 50 m 0.53 mm 1 μm Each
10216611 60 m 0.25 mm 0.25 μm Each
10718242 60 m 0.32 mm 0.25 μm Each
10348021 60 m 0.32 mm 0.5 μm Each
27 options
Product Code. 10727472
Brand: Macherey-Nagel™ 723555.25
Only null left
Specifications
Diameter (Metric) 0.8 mm
Type GC Capillary Column
Diameter (Metric) Inner 0.53 mm
Stationary Phase Permabond™ FFAP
USP Type G25, G35
For Use With (Application) Free Carboxylic Acids Analysis, Free Carboxylic Acids
Temperature Range 220°C to 240°C
Film Thickness 1 μm
Length (Metric) 25 m
Packing Material Permabond™ FFAP
Quantity 1 Each