Learn More
- Home
- Products
- Chromatography Columns and Cartridges
- GC Columns
- GC Capillary Columns
- Macherey-Nagel™ Permabond™ FFAP GC Capillary Column
Macherey-Nagel™ Permabond™ FFAP GC Capillary Column
Product Code. 10586501 Shop All Macherey-Nagel Products
Change view
Click to view available options
Length (Metric):
10 m
20 m
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.1 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.1 μm
0.25 μm
0.35 μm
0.5 μm
1 μm
Unit Size:
Each
Pack of 10
| Product Code. | Length (Metric) | Diameter (Metric) Inner | Film Thickness | unitSize |
|---|---|---|---|---|
| 10586501 | 10 m | 0.1 mm | 0.25 μm | Each |
| 10786122 | 10 m | 0.1 mm | 0.1 μm | Each |
| 10601112 | 10 m | 0.25 mm | 0.25 μm | Pack of 10 |
| 10033011 | 10 m | 0.32 mm | 0.35 μm | Each |
| 10348641 | 10 m | 0.32 mm | 0.5 μm | Each |
| 10398301 | 10 m | 0.53 mm | 1 μm | Each |
| 10236801 | 20 m | 0.1 mm | 0.1 μm | Each |
| 10698801 | 25 m | 0.25 mm | 0.1 μm | Each |
| 10368541 | 25 m | 0.25 mm | 0.25 μm | Each |
| 10440512 | 25 m | 0.32 mm | 0.1 μm | Each |
| 10556341 | 25 m | 0.32 mm | 0.25 μm | Each |
| 10318641 | 25 m | 0.32 mm | 0.35 μm | Each |
| 10576861 | 25 m | 0.32 mm | 0.5 μm | Each |
| 10727472 | 25 m | 0.53 mm | 1 μm | Each |
| 10479931 | 30 m | 0.25 mm | 0.25 μm | Each |
| 10469741 | 30 m | 0.32 mm | 0.25 μm | Each |
| 10338021 | 30 m | 0.32 mm | 0.5 μm | Each |
| 10023011 | 50 m | 0.25 mm | 0.25 μm | Each |
| 10796122 | 50 m | 0.25 mm | 0.1 μm | Each |
| 10708242 | 50 m | 0.32 mm | 0.25 μm | Each |
| 10092851 | 50 m | 0.32 mm | 0.35 μm | Each |
| 10348211 | 50 m | 0.32 mm | 0.1 μm | Each |
| 10065841 | 50 m | 0.32 mm | 0.5 μm | Each |
| 10072961 | 50 m | 0.53 mm | 1 μm | Each |
| 10216611 | 60 m | 0.25 mm | 0.25 μm | Each |
| 10718242 | 60 m | 0.32 mm | 0.25 μm | Each |
| 10348021 | 60 m | 0.32 mm | 0.5 μm | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Specifications
Specifications
| Diameter (Metric) | 0.4 mm |
| Type | GC Capillary Column |
| Diameter (Metric) Inner | 0.1 mm |
| Stationary Phase | Permabond™ FFAP |
| USP Type | G25, G35 |
| For Use With (Application) | Free Carboxylic Acids Analysis, Free Carboxylic Acids |
| Temperature Range | 220°C to 240°C |
| Film Thickness | 0.25 μm |
| Length (Metric) | 10 m |
| Packing Material | Permabond™ FFAP |
| Quantity | 1 Each |
| Show More Show Less |