Learn More
Macherey-Nagel™ Permabond™ FFAP GC Capillary Column
Shop All Macherey-Nagel Products
Click to view available options
Length (Metric):
10 m
20 m
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.1 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.1 μm
0.25 μm
0.35 μm
0.5 μm
1 μm
Unit Size:
Each
Pack of 10
Specifications
Specifications
| Diameter (Metric) | 0.8 mm |
| Type | GC Capillary Column |
| Diameter (Metric) Inner | 0.53 mm |
| Stationary Phase | Permabond™ FFAP |
| USP Type | G25, G35 |
| For Use With (Application) | Free Carboxylic Acids Analysis, Free Carboxylic Acids |
| Temperature Range | 220°C to 240°C |
| Film Thickness | 1 μm |
| Length (Metric) | 10 m |
| Packing Material | Permabond™ FFAP |
| Show More |