Learn More
Trajan Scientific and Medical™ PerkinElmer Series 200 LC Autosampler Syringe
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for PerkinElmer instruments
Specifications
Specifications
| Material | PTFE (Plunger) |
| For Use With (Application) | High-performance Liquid Chromatography |
| Volume (Metric) | 250 μL |
| Connection Type | Gas Tight |