Learn More
Trajan Scientific and Medical™ PerkinElmer Series 200 LC Autosampler Syringe
Product Code. 11947224
Change view
Click to view available options
Volume (Metric):
50 μL
100 μL
250 μL
500 μL
1 mL
Unit Size:
Each
Product Code. Volume (Metric) unitSize
11947224 500 μL Each
11927224 100 μL Each
12760710 1 mL Each
12740710 250 μL Each
11917224 50 μL Each
5 options
Product Code. 11947224
Brand: Trajan Scientific and Medical™ 007995
Accurate and reproducible injections.
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for PerkinElmer instruments
Specifications
Material PTFE (Plunger)
For Use With (Application) High-performance Liquid Chromatography
Volume (Metric) 500 μL
Connection Type Gas Tight