missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Peripherin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA316723
3401.25 NOK valid until 2025-12-19
BEST PRICE on Promo! Use promo code "24110" to get your promotional price.
This item is not returnable.
View return policy
Description
Peripherin Polyclonal Antibody for Western Blot, IHC (P), IHC (F), ICC/IF
Peripherin is a 57kD type III intermediate filament that is a specific marker for peripheral neurons, including enteric ganglion cells. Peripherin is expressed in the developing peripheral nervous system and is highly enriched in neuronal derivatives of the neural crest. Peripherin offers an advantage over other neural markers, such as S100 and NSE in that it does not stain chrmaffin cell types.
Specifications
| Peripherin | |
| Polyclonal | |
| Unconjugated | |
| PRPH | |
| if3; nef4; Neurofilament 4; neurofilament 4 (57kD); neuronal intermediate filament IF3; Perf; Peripherin; peripherin 1; peripherin L homeolog; plasticin; PRPH; prph.L; PRPH1; XELAEV_18013332mg; XIF3 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Bovine, Pig | |
| Antibody | |
| IgG |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Conc. Not Determined | |
| Whole serum with 0.035% sodium azide | |
| A6QQJ3, P15331, P21807, P41219 | |
| PRPH | |
| Full length recombinant human peripherin expressed in and purified from E. coli. | |
| RUO | |
| 100152434, 19132, 24688, 510082, 5630 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction