Learn More
Perindoprilat, TRC
Product Code. 18072350
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18072350 5 mg 5mg
18092340 10 mg 10mg
18060350 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18072350 Supplier Toronto Research Chemicals Supplier No. TRCP2875355MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 95153-31-4
Specifications
Purity >95% (HPLC)
Format Neat
Molecular Formula C17H28N2O5
MDL Number MFCD00865777
Molecular Weight (g/mol) 340.41, 340.42
InChI Key ODAIHABQVKJNIY-PEDHHIEDSA-N
SMILES CCC[C@H](N[C@@H](C)C(=O)N1[C@H]2CCCC[C@H]2C[C@H]1C(O)=O)C(O)=O
IUPAC Name (2S,3aS,7aS)-1-[(2S)-2-{[(1S)-1-carboxybutyl]amino}propanoyl]-octahydro-1H-indole-2-carboxylic acid
Product Type Metabolite∣Impurity
Quantity 5 mg
Shipping Condition Room Temperature
Synonym 1H-Indole-2-carboxylic acid, 1-[(2S)-2-[[(1S)-1-carboxybutyl]amino]-1-oxopropyl]octahydro-, (2S,3aS,7aS)-∣1H-Indole-2-carboxylic acid, 1-[2-[(1-carboxybutyl)amino]-1-oxopropyl]octahydro-, [2S-[1[R*(R*)],2α,3aβ,7aβ]]-∣Perindoprilat∣S 9780∣Perindopril RC B∣Perindopril desethyl