missing translation for 'onlineSavingsMsg'
Learn More

Perilipin Antibody, PerCP, Novus Biologicals™

Product Code. 18568501 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
Product Code. Quantity unitSize
18568501 0.1 mL 0.10mL
1 options
This item is not returnable. View return policy

Product Code. 18568501

Brand: Novus Biologicals NB11040760PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Perilipin Polyclonal antibody specifically detects Perilipin in Human, Mouse, Rat, Porcine, Fish samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Perilipin
Applications Flow Cytometry
Classification Polyclonal
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS
Gene Alias PERI, perilipin, perilipin 1, perilipin-1, PLINLipid droplet-associated protein
Host Species Rabbit
Immunogen This Perilipin Antibody was developed against a synthetic peptide made to a region between residues 450-522 (C-terminus) of the human Perilipin protein. [Swiss-Prot# O60240]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism, Lipid Droplets
Primary or Secondary Primary
Gene ID (Entrez) 5346
Target Species Human, Mouse, Rat, Pig, Fish
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.