missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pericentrin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
2075.00 NOK - 2985.00 NOK
Specifications
| Antigen | Pericentrin |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 1:100-1:500 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18649641
|
Novus Biologicals
NBP2-97596-100ul |
100 μg |
2985.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654614
|
Novus Biologicals
NBP2-97596-50ul |
50 μg |
2075.00 NOK
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pericentrin Polyclonal antibody specifically detects Pericentrin in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| Pericentrin | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, Centrosome Markers | |
| PBS (pH 7.0) | |
| 5116 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Kendrin, KIAA0402, MOPD2, PCN, PCNT2pericentrin B, PCNTB, pericentrin, pericentrin 2 (kendrin), pericentrin, kendrin (KIAA0402)10KENPCTN2, pericentrin-2, pericentrin-380, pericentrin-B, SCKL4kendrin | |
| Produced in rabbits immunized with E. coli-derived Human Pericentrin fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title