Learn More
BINDER™ Perforated Shelf Rack
Product Code. 30238292 Shop All BINDER Products
Change view
Click to view available options
For Use With (Equipment):
KB/KB PRO 1060, KBF/KBF PRO 1060
KB/KB PRO 130, KBF/KBF PRO 130
KB/KB PRO 720, KBF/KBF PRO 720
KB/KB PRO/KBF/KBF PRO 260/470
KBF/KBF PRO 1600, KB PRO 1600
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
30238292 KB/KB PRO 1060, KBF/KBF PRO 1060 Each
30238348 KB/KB PRO 130, KBF/KBF PRO 130 Each
30238205 KB/KB PRO/KBF/KBF PRO 260/470 Each
30238227 KB/KB PRO 720, KBF/KBF PRO 720 Each
30238250 KBF/KBF PRO 1600, KB PRO 1600 Each
5 options
Product Code. 30238292
Brand: BINDER™ 80122409
Only null left
Add to basket
Add to basket
Perforated shelf rack in-set with pair of retaining U-rails, for use with climate chambers and cooling incubator chambers.
  • Shelf rack made of stainless steel
  • Includes set of U-rails
Specifications
For Use With (Equipment) KB/KB PRO 1060, KBF/KBF PRO 1060
Material Stainless Steel
Type Rack