Learn More
Perfluorophenyl acetate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 19220-93-0 |
| Molecular Formula | C8H3F5O2 |
| MDL Number | MFCD00015552 |
| Quantity | 25 g |
| SMILES | CC(OC1=C(F)C(F)=C(F)C(F)=C1F)=O |
| Molecular Weight (g/mol) | 226.10 |
| Percent Purity | 98% |
| Chemical Name or Material | Perfluorophenyl acetate |