Learn More
Perfluoro-compound FC-40(TM), Thermo Scientific Chemicals
Product Code. 10173360 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
250 g
Packaging:
Glass bottle
Unit Size:
100g
10g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10173360 250 g Glass bottle 250g
10163360 10 g Glass bottle 10g
10081150 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10173360 Supplier Thermo Scientific Acros Supplier No. 123762500
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 51142-49-5
Molecular Formula C21F48N2
Molecular Weight (g/mol) 1192.168
MDL Number MFCD01632188
InChI Key QDOIZVITZUBGOQ-UHFFFAOYSA-N
Synonym fluorinert fc-40, perfluoro-compound fc-40, bis nonafluorobutyl trifluoromethyl amine; perfluorotributylamine, 1,1,2,2,3,3,4,4,4-nonafluoro-n,n-bis 1,1,2,2,3,3,4,4,4-nonafluorobutyl butan-1-amine,1,1,2,2,3,3,4,4,4-nonafluoro-n-1,1,2,2,3,3,4,4,4-nonafluorobutyl-n-trifluoromethyl butan-1-amine, 1,1,2,2,3,3,4,4,4-nonafluoro-n,n-bis 1,1,2,2,3,3,4,4,4-nonafluorobutyl butan-1-amine; 1,1,2,2,3,3,4,4,4-nonafluoro-n-1,1,2,2,3,3,4,4,4-nonafluorobutyl-n-trifluoromethyl butan-1-amine, bis 1,1,2,2,3,3,4,4,4-nonafluorobutyl trifluoromethyl amine; perfluorotributylamine
PubChem CID 2723673
IUPAC Name 1,1,2,2,3,3,4,4,4-nonafluoro-N,N-bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)butan-1-amine;1,1,2,2,3,3,4,4,4-nonafluoro-N-(1,1,2,2,3,3,4,4,4-nonafluorobutyl)-N-(trifluoromethyl)butan-1-amine
SMILES C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F.C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(F)(F)F)(F)F)(F)F
Specifications
Color Colorless
Density 1.8700g/mL
Boiling Point 158°C to 173°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 250 g
Viscosity 1.6-2.8cSt (25°C)
Formula Weight 650
Physical Form Liquid
Chemical Name or Material Perfluoro-compound FC-40™
Product Identifier
  • Perfluoro-compound FC-40]TS
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
Hazard Statement
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement