Learn More
Perfluoro(2-methyl-3-pentanone), TRC
Product Code. 30418662
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30418662 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30418662 Supplier Toronto Research Chemicals Supplier No. TRCP999235500MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Perfluoro-2-methyl-3-pentanone
CAS 756-13-8
Molecular Formula C6 F12 O
Quantity 500 mg
Synonym 1,1,1,2,2,4,5,5,5-Nonafluoro-4-(trifluoromethyl)-3-pentanone, Dodecafluoro-2-methylpentan-3-one, Heptafluoroisopropyl pentafluoroethyl ketone, Khaldon PFK 49, L 18543, Novec 1230, Novec 649, Perfluoro(ethyl isopropyl ketone), Perfluoro-(4-methylpentan-3-one), Perfluoro-2-methyl-3-pentanone
InChI Formula InChI=1S/C6F12O/c7-2(4(10,11)12,5(13,14)15)1(19)3(8,9)6(16,17)18
SMILES FC(F)(F)C(F)(F)C(=O)C(F)(C(F)(F)F)C(F)(F)F
IUPAC Name 1,1,1,2,2,4,5,5,5-nonafluoro-4-(trifluoromethyl)pentan-3-one
Molecular Weight (g/mol) 316.04
Formula Weight 315.9758