Learn More
Pepsin, Extra Pure, Powder, SLR, Fisher Chemical™
Product Code. p-7073369 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. Quantity unitSize
11428712 25 g 25g
10264440 100 g 100g
10386140 500 g 500g
3 options
Product Code. 11428712
Brand: Fisher Chemical P/1120/46
Request Bulk or Custom Format
Enzyme that breaks down proteins into smaller peptides.
Proteolitic activity BPC: 2500 - 3000 (Based on the British Pharmaceutical Codex 1959 (BPC) for the pepsin proteolytic activity report. Dissolves not less than 2500 times its weight of coagulated egg albumin.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 25 g
Product Type Pepsin
Form Powder
compliance-icons compliance-icons
Product Identifier
  • Pepsin
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.