Learn More
Invitrogen™ Pepsin A Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5103156
Product Code. 16699604
  • 4705.00 NOK / 100µL
Description
Antibody detects endogenous levels of total Pepsin A.
Pepsin consists of a single polypeptide chain and arises from its precursor, pepsinogen, by removal of a 41 amino acid segment from the N-terminus. Pepsinogen is synthesized in the stomach lining, and hydrochloric acid, also produced by the gastric mucosa, is necessary to convert the inactive enzyme and to maintain the optimum acidity (pH 1-3) for pepsin function. Pepsin is particularly effective in cleaving peptide bonds involving aromatic amino acids. Pepsin shows extremely broad specificity; although bonds involving phenylalanine and leucine are preferred, many others are also cleaved to some extent. Pepsin A is a member of the subfamily A1 within the pepsin family and is the predominant endopeptidase in the gastric juice of vertebrates. Pepsin A is inhibited by ovUS-1, a uterine serpin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pepsin A
Polyclonal
Unconjugated
PGA3
1110035E17Rik; Pepf; pepsin A; pepsin A-3; Pepsin A-4; pepsin A-5; Pepsin F; pepsinogen 3, group I (pepsinogen A); pepsinogen 4, group I (pepsinogen A); pepsinogen 5, group I; pepsinogen 5, group I (pepsinogen A); pepsinogen A3; pepsinogen A4; pepsinogen A5; pepsinogen F; pepsinogen F protein; pepsinogen-3; Pepsinogen-4; pepsinogen-5; Pg5; PGA3; PGA4; PGA5
Rabbit
Affinity Chromatography
RUO
5222, 58803, 643834, 643847
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P0DJD7, P0DJD8, P0DJD9, Q9D106
PGA3, PGA4, PGA5
A synthesized peptide derived from human PGA4(Accession P0DJD7), corresponding to amino acid residues N352-A388.
100 μL
Primary
Human, Mouse