Learn More
Invitrogen™ Pepsin A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103156
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Pepsin A.
Pepsin consists of a single polypeptide chain and arises from its precursor, pepsinogen, by removal of a 41 amino acid segment from the N-terminus. Pepsinogen is synthesized in the stomach lining, and hydrochloric acid, also produced by the gastric mucosa, is necessary to convert the inactive enzyme and to maintain the optimum acidity (pH 1-3) for pepsin function. Pepsin is particularly effective in cleaving peptide bonds involving aromatic amino acids. Pepsin shows extremely broad specificity; although bonds involving phenylalanine and leucine are preferred, many others are also cleaved to some extent. Pepsin A is a member of the subfamily A1 within the pepsin family and is the predominant endopeptidase in the gastric juice of vertebrates. Pepsin A is inhibited by ovUS-1, a uterine serpin.
Specifications
| Pepsin A | |
| Polyclonal | |
| Unconjugated | |
| PGA3 | |
| 1110035E17Rik; Pepf; pepsin A; pepsin A-3; Pepsin A-4; pepsin A-5; Pepsin F; pepsinogen 3, group I (pepsinogen A); pepsinogen 4, group I (pepsinogen A); pepsinogen 5, group I; pepsinogen 5, group I (pepsinogen A); pepsinogen A3; pepsinogen A4; pepsinogen A5; pepsinogen F; pepsinogen F protein; pepsinogen-3; Pepsinogen-4; pepsinogen-5; Pg5; PGA3; PGA4; PGA5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 5222, 58803, 643834, 643847 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P0DJD7, P0DJD8, P0DJD9, Q9D106 | |
| PGA3, PGA4, PGA5 | |
| A synthesized peptide derived from human PGA4(Accession P0DJD7), corresponding to amino acid residues N352-A388. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |