Learn More
PEO-IAA, MedChemExpress
Product Code. 16414698
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 16414698
Brand: Medchem Express HY112730
MedChemExpress PEO-IAA is an indole-3-acetic acid (IAA) antagonist. PEO-IAA is an auxin antagonist that binds to transport inhibitor response 1/auxin signaling F-box proteins (TIR1/AFBs).
PEO-IAA is an indole-3-acetic acid (IAA) antagonist. PEO-IAA is an auxin antagonist that binds to transport inhibitor response 1/auxin signaling F-box proteins (TIR1/AFBs).
Specifications
Chemical Name or Material PEO-IAA
Physical Form Solid
CAS 6266-66-6
Color Light Nude
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C18H15NO3
Synonym 2-(1H-Indol-3-yl)-4-oxo-4-phenyl-butyric acid
Solubility Information DMSO : 50 mg/mL (170.46 mM; Need ultrasonic)
SMILES O=C(O)C(CC(C1=CC=CC=C1)=O)C2=CNC3=C2C=CC=C3
Molecular Weight (g/mol) 293.32
Formula Weight 293.32
Percent Purity 98.08%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.