Learn More
Pentoxifylline, Mikromol™
Description
Pentoxifylline
Specifications
Specifications
| Chemical Name or Material | Pentoxifylline |
| CAS | 5-6-6493 |
| Analyte or Component Names | Pentoxifylline |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H18 N4 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 1-(5-Oxohexyl)-3,7-dimethylxanthine, 1-(5-Oxohexyl)theobromine, 3,7-Dihydro-3,7-dimethyl-1-(5-oxohexyl)-1H-purine-2,6-dione, 3,7-Dimethyl-1-(5-oxohexyl)-1H,3H-purin-2,6-dione, 3,7-Dimethyl-1-(5-oxohexyl)xanthine, Agapurin, Agapurin Retard, Azupentat, BL 191, Dimethyloxohexylxanthine, Durapental, NSC 637086, Oxpentifylline, PTX, Pentoxifyllin, Pentoxifylline, Pentoxiphyllin, Pentoxiphylline, Pentoxyfilline, Pentoxyphyllin, Pexal, Rentylin, Torental, Trental, Vasotal, Vazofirin |
| InChI Formula | InChI=1S/C13H18N4O3/c1-9(18)6-4-5-7-17-12(19)10-11(14-8-15(10)2)16(3)13(17)20/h8H,4-7H2,1-3H3 |
| SMILES | CN1C(=O)N(CCCCC(=O)C)C(=O)c2c1ncn2C |
| IUPAC Name | 3,7-dimethyl-1-(5-oxohexyl)purine-2,6-dione |
| Show More |