Learn More
Pentostatin, TRC
Product Code. 30426204
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426204 5 mg 5mg
30412106 10 mg -
30413122 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30426204 Supplier Toronto Research Chemicals Supplier No. TRCP2273505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 53910-25-1
Molecular Formula C11 H16 N4 O4
Molecular Weight (g/mol) 268.27
Synonym (8R)-3-(2-Deoxy-beta-D-erythro-pentofuranosyl)-3,4,7,8-tetrahydro-imidazo[4,5-d][1,3]diazepin-8-ol, (R)-, 2'-DCF, 2'-Deoxycoformycin, CI 825, CL 67310465, Cl 825, Co-V, Co-Vidarabine, Coforin, NSC 218321, NSC 247520, Nipent, PD-ADI, Vira A Deaminase Inhibitor
IUPAC Name (8R)-3-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-7,8-dihydro-6H-imidazo[4,5-d][1,3]diazepin-8-ol
SMILES OCC1OC(N2C=NC3=C2NC=NCC3O)CC1O
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C11H16N4O4/c16-3-8-6(17)1-9(19-8)15-5-14-10-7(18)2-12-4-13-11(10)15/h4-9,16-18H,1-3H2,(H,12,13)/t6-,7+,8+,9+/m0/s1
Formula Weight 268.1172
Chemical Name or Material Pentostatin