missing translation for 'onlineSavingsMsg'
Learn More

Pentostatin, MedChemExpress

Product Code. p-200098906
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
missing translation for 'unitSize'
10mg
50mg
This item is not returnable. View return policy

Product Code. 16448258

missing translation for 'mfr': Medchem Express HYA0006

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pentostatin (CI-825; Deoxycoformycin) is an irreversible inhibitor of adenosine deaminase with Ki of 2.5 pM.

Pentostatin (CI-825

Specifications

Chemical Name or Material Pentostatin
CAS 53910-25-1
Color White
Purity Grade Notes Research
Molecular Formula C11H16N4O4
Quantity 10 mg
Synonym CI-825 Deoxycoformycin
Solubility Information H2O : 100 mg/mL (372.76 mM; Need ultrasonic) ∣DMSO : ≥ 50 mg/mL (186.38 mM)
SMILES O[C@H]1C(N=CN2[C@H](O[C@@H]3CO)C[C@@H]3O)=C2NC=NC1
Molecular Weight (g/mol) 268.27
Formula Weight 268.27
Percent Purity 95.54%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.