Learn More
Pentostatin, MedChemExpress
Product Code. 16448258
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16448258 10 mg 10mg
16424147 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16448258 Supplier Medchem Express Supplier No. HYA0006
MedChemExpress Pentostatin (CI-825; Deoxycoformycin) is an irreversible inhibitor of adenosine deaminase with Ki of 2.5 pM.
Pentostatin (CI-825
Specifications
Chemical Name or Material Pentostatin
CAS 53910-25-1
Color White
Purity Grade Notes Research
Molecular Formula C11H16N4O4
Quantity 10 mg
Synonym CI-825 Deoxycoformycin
Solubility Information H2O : 100 mg/mL (372.76 mM; Need ultrasonic) ∣DMSO : ≥ 50 mg/mL (186.38 mM)
SMILES O[C@H]1C(N=CN2[C@H](O[C@@H]3CO)C[C@@H]3O)=C2NC=NC1
Molecular Weight (g/mol) 268.27
Formula Weight 268.27
Percent Purity 95.54%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death