Learn More
Pentanoic-2,2-d2 Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Pentanoic-2,2-d2 Acid |
| CAS | 64118-37-2 |
| Molecular Formula | C5 2H2 H8 O2 |
| Quantity | 0.5 g |
| InChI Formula | InChI=1 S/C5H10O2/c1-2-3-4-5(6)7/h2-4H2,1H3,(H,6,7)/i4D2 |
| SMILES | [2 H]C([2 H])(CCC)C(=O)O |
| IUPAC Name | 2,2-dideuteriopentanoic acid |
| Molecular Weight (g/mol) | 104.144 |
| Formula Weight | 104.081 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |