missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentakis(diethylamino)niobium(V), 99.9% (metals basis), mixture of complexes, Thermo Scientific™
Click to view available options
Quantity:
500 mg
2.5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Boiling Point | 120°C (0.1 mmHg) |
Odor | Amine-like |
Linear Formula | Nb[N(C2H5)2]5 |
Quantity | 500 mg |
UN Number | UN3267 |
Sensitivity | Moisture sensitive |
Formula Weight | 453.56 |
Percent Purity | 99.9% |
Physical Form | Liquid |
Assay | (metals basis) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'