missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 4-(tetrahydropyran-4-yloxy)benzoate, 97%, Thermo Scientific™

Product Code. 10727865
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10727865

Brand: Thermo Scientific Maybridge CC52426DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 930110-97-7
Molecular Formula C18H13F5O4
Molecular Weight (g/mol) 388.29
MDL Number MFCD09702368
InChI Key BDWCEFHYQPZQRO-UHFFFAOYSA-N
Synonym pentafluorophenyl 4-tetrahydropyran-4-yloxy benzoate, pentafluorophenyl 4-oxan-4-yloxy benzoate, pentafluorophenyl 4-oxan-4-yl oxy benzoate, 2,3,4,5,6-pentafluorophenyl 4-oxan-4-yloxy benzoate, 2,3,4,5,6-pentakis fluoranyl phenyl 4-oxan-4-yloxy benzoate, 4-4-oxanyloxy benzoic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 24229528
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 4-(oxan-4-yloxy)benzoate
SMILES FC1=C(F)C(F)=C(OC(=O)C2=CC=C(OC3CCOCC3)C=C2)C(F)=C1F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 388.29
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl 4-(tetrahydropyran-4-yloxy)benzoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.