Learn More
Pentafluorophenyl 4-morpholinobenzoate, 97%, Thermo Scientific™
Product Code. 10695125
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10695125 1 g Amber Glass Bottle 1g
10537634 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10695125 Supplier Thermo Scientific Maybridge Supplier No. CC17426DA
 Restricted Item
Estimated Shipment: 04-12-2026
Chemical Identifiers
CAS 921938-51-4
Molecular Formula C17H12F5NO3
Molecular Weight (g/mol) 373.28
MDL Number MFCD09817462
InChI Key UJYQDNSLPIRXRT-UHFFFAOYSA-N
Synonym pentafluorophenyl 4-morpholin-4-ylbenzoate, pentafluorophenyl 4-morpholinobenzoate, pentafluorophenyl 4-morpholin-4-yl benzoate, 2,3,4,5,6-pentafluorophenyl 4-morpholin-4-yl benzoate, perfluorophenyl 4-morpholinobenzoate, 2,3,4,5,6-pentakis fluoranyl phenyl 4-morpholin-4-ylbenzoate, 4-4-morpholinyl benzoic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 24229462
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 4-morpholin-4-ylbenzoate
SMILES FC1=C(F)C(F)=C(OC(=O)C2=CC=C(C=C2)N2CCOCC2)C(F)=C1F
Specifications
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 373.28
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl 4-morpholinobenzoate