Learn More
Pentafluorophenyl 2-morpholino-1,3-thiazole-5-carboxylate, 97%, Thermo Scientific™
Product Code. p-3586884
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Product Code. 10205604
Brand: Thermo Scientific Maybridge CC62426CB
Request Bulk or Custom Format
Chemical Identifiers
CAS 941716-88-7
Molecular Formula C14H9F5N2O3S
Molecular Weight (g/mol) 380.289
MDL Number MFCD09966152
InChI Key ZFWIQYGUHPHWEU-UHFFFAOYSA-N
Synonym pentafluorophenyl 2-morpholino-1,3-thiazole-5-carboxylate, pentafluorophenyl 2-morpholin-4-yl-1,3-thiazole-5-carboxylate, 2,3,4,5,6-pentafluorophenyl 2-morpholin-4-yl-1,3-thiazole-5-carboxylate, 2,3,4,5,6-pentakis fluoranyl phenyl 2-morpholin-4-yl-1,3-thiazole-5-carboxylate, 2-4-morpholinyl-5-thiazolecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 43811041
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 2-morpholin-4-yl-1,3-thiazole-5-carboxylate
SMILES C1COCCN1C2=NC=C(S2)C(=O)OC3=C(C(=C(C(=C3F)F)F)F)F
Specifications
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 380.29
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl 2-morpholino-1, 3-thiazole-5-carboxylate