Learn More
Pentafluorobenzaldehyde, 98%
Description
Pentafluorobenzaldehyde was used as derivatization reagent in a study to develop a sensitive method for the determination of primary amines in sewage sludge by headspace solid-phase micro extraction and gas chromatography-tandem mass spectrometry. It was used in the synthesis of novel, high molecular weight fluorinated aromatic polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Pentafluorobenzaldehyde was used as derivatization reagent in a study to develop a sensitive method for the determination of primary amines in sewage sludge by headspace solid-phase micro extraction and gas chromatography-tandem mass spectrometry. It was used in the synthesis of novel, high molecular weight fluorinated aromatic polymers.
Solubility
Sparingly Soluble in water (0.027 g/L at 25°C).
Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent, air, bases.
Specifications
Specifications
| Melting Point | 24°C to 27°C |
| Density | 1.63 |
| Boiling Point | 164°C to 166°C |
| Flash Point | 77°C (170°F) |
| Refractive Index | 1.451 |
| Quantity | 25 g |
| UN Number | UN2811 |
| Beilstein | 1876550 |
| Sensitivity | Air Sensitive |
| Solubility Information | Sparingly Soluble in water (0.027g/L at 25°C). |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.