Learn More
Penoxsulam, TRC
Product Code. 30004299
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004299 250 mg 250mg
30011346 500 mg 500mg
17940735 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30004299 Supplier Toronto Research Chemicals Supplier No. TRCP274000250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 219714-96-2
Molecular Formula C16 H14 F5 N5 O5 S
Molecular Weight (g/mol) 483.37
Synonym 2-(2,2-Difluoroethoxy)-N-(5,8-dimethoxy[1,2,4]triazolo[1,5-c]pyrimidin-2-yl)-6-(trifluoromethyl)benzenesulfonamide, Clipper, Clipper (benzenesulfonamide), Clipper 20OD, DASH 001, Granite, Granite (herbicide), Granite SC, Grasp, Grasp (benzenesulfonamide), Grasp EC, Penoxsulam, Rainbow 25OD, Ricer, Ricer 240SC, Viper
IUPAC Name 2-(2,2-difluoroethoxy)-N-(5,8-dimethoxy-[1,2,4]triazolo[1,5-c]pyrimidin-2-yl)-6-(trifluoromethyl)benzenesulfonamide
SMILES COc1cnc(OC)n2nc(NS(=O)(=O)c3c(OCC(F)F)cccc3C(F)(F)F)nc12
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C16H14F5N5O5S/c1-29-10-6-22-15(30-2)26-13(10)23-14(24-26)25-32(27,28)12-8(16(19,20)21)4-3-5-9(12)31-7-11(17)18/h3-6,11H,7H2,1-2H3,(H,24,25)
Formula Weight 483.0636
Percent Purity >95%
Chemical Name or Material Penoxsulam
Grade HPLC