Learn More
Penoxsulam 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30325074
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30325074 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30325074 Supplier Dr. Ehrenstorfer Supplier No. DREXA15937000AL
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Penoxsulam
CAS 219714-96-2
Analyte or Component Names Penoxsulam
Shipping Condition Room Temperature
Molecular Formula C16 H14 F5 N5 O5 S
Concentration 100 μg/mL
Certifications/Compliance ISO/IEC 17025
Synonym 2-(2,2-Difluoroethoxy)-N-(5,8-dimethoxy[1,2,4]triazolo[1,5-c]pyrimidin-2-yl)-6-(trifluoromethyl)benzenesulfonamide, Clipper, Clipper (benzenesulfonamide), Clipper 20OD, DASH 001, Granite, Granite (herbicide), Granite SC, Grasp, Grasp (benzenesulfonamide), Grasp EC, Penoxsulam, Rainbow 25OD, Ricer, Ricer 240SC, Viper
InChI Formula InChI=1S/C16H14F5N5O5S/c1-29-10-6-22-15(30-2)26-13(10)23-14(24-26)25-32(27,28)12-8(16(19,20)21)4-3-5-9(12)31-7-11(17)18/h3-6,11H,7H2,1-2H3,(H,24,25)
Solution Type Single Solution
SMILES COc1cnc(OC)n2nc(NS(=O)(=O)c3c(OCC(F)F)cccc3C(F)(F)F)nc12
IUPAC Name 2-(2,2-difluoroethoxy)-N-(5,8-dimethoxy-[1,2,4]triazolo[1,5-c]pyrimidin-2-yl)-6-(trifluoromethyl)benzenesulfonamide
Solvent or Matrix Acetonitrile
Industry Type Environmental Testing
Molecular Weight (g/mol) 483.37
Quantity 1 mL
Formula Weight 483.0636 g/mol