Learn More
Penicillin V Potassium, MedChemExpress
Product Code. 16418546
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 16418546
Brand: Medchem Express HYB0975
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress Penicillin V Potassium (Phenoxymethylpenicillin potassium salt) is an orally active antibiotic. Penicillin V Potassium inhibits the growth of Streptococci, C. difficile and S. aureus. Penicillin V Potassium can be used for the research of otitis, sinusitis, pharyngitis and tonsillitis.
Penicillin V Potassium (Phenoxymethylpenicillin potassium salt) is an orally active antibiotic. Penicillin V Potassium inhibits the growth of Streptococci, C. difficile and S. aureus. Penicillin V Potassium can be used for the research of otitis, sinusitis, pharyngitis and tonsillitis.
Specifications
Chemical Name or Material Penicillin V Potassium
Physical Form Solid
CAS 132-98-9
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C16H17KN2O5S
For Use With (Application) COVID-19-immunoregulation
Synonym Phenoxymethylpenicillin potassium salt
Solubility Information DMSO : 12 mg/mL (30.89 mM; Need ultrasonic) ∣H2O : 6 mg/mL (15.44 mM; Need ultrasonic and warming)
SMILES O=C(O[K])[C@@H](C(C)(C)S[C@]1([H])[C@@H]2NC(COC3=CC=CC=C3)=O)N1C2=O
Molecular Weight (g/mol) 388.48
Formula Weight 388.48
Percent Purity 99.02%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.