missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Penicillin Monoclonal Antibody (P2B9)
GREENER_CHOICE

Product Code. 11355173
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Product Code. Quantity unitSize
11355173 100 μg 100µg
1 options
This item is not returnable. View return policy

Product Code. 11355173

Brand: Invitrogen™ MA514714

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA5-14714 targets Penicillin in ELISA, IP, and RIA applications and shows reactivity with Chemical samples. The MA5-14714 immunogen is penicillin-BSA. Product MA514714 is a smaller package size of MIP0701 (formerly sold as a Seradyn product).

Penicillin (sometimes abbreviated PCN or pen) is a group of antibiotics derived from Penicillium fungi. Penicillin antibiotics are historically significant because they are the first drugs that were effective against many previously serious diseases such as syphilis and Staphylococcus infections. Penicillins are still widely used today, though many types of bacteria are now resistant. All penicillins are Beta-lactam antibiotics and are used in the treatment of bacterial infections caused by susceptible, usually Gram-positive, organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Penicillin
Applications ELISA, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone P2B9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
Gene Alias PCN
Host Species Mouse
Immunogen Penicillin-BSA.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.