Learn More
Penfluridol, Mikromol™
Description
Penfluridol
Specifications
Specifications
| Chemical Name or Material | Penfluridol |
| CAS | 26864-56-2 |
| Analyte or Component Names | Penfluridol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C28 H27 Cl F5 N O |
| Certifications/Compliance | ISO 17025 |
| Synonym | Penfluridol, 1-[4,4-Bis(4-fluorophenyl)butyl]-4-[4-chloro-3-(trifluoromethyl)phenyl]-4-piperidinol, 1-[4,4-bis(p-fluorophenyl)butyl]-4-(4-chloro-α,α,α-trifluoro-m-tolyl)-4-piperidinol, 1-(4,4-bis(4-fluorophenyl)butyl)-4-hydroxy-4-(3-trifluoromethyl-4-chlorophenyl)piperidine, R-16341 |
| InChI Formula | InChI=1S/C28H27ClF5NO/c29-26-12-7-21(18-25(26)28(32,33)34)27(36)13-16-35(17-14-27)15-1-2-24(19-3-8-22(30)9-4-19)20-5-10-23(31)11-6-20/h3-12,18,24,36H,1-2,13-17H2 |
| SMILES | OC1(CCN(CCCC(c2ccc(F)cc2)c3ccc(F)cc3)CC1)c4ccc(Cl)c(c4)C(F)(F)F |
| IUPAC Name | 1-[4,4-bis(4-fluorophenyl)butyl]-4-[4-chloro-3-(trifluoromethyl)phenyl]piperidin-4-ol |
| Show More |