missing translation for 'onlineSavingsMsg'
Learn More

Penfluridol, MedChemExpress

Product Code. p-200099387
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494537 50 mg 50mg
16418596 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16494537 Supplier Medchem Express Supplier No. HYB1077

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Penfluridol is a highly potent, first generation diphenylbutylpiperidine antipsychotic.

Penfluridol is a highly potent, first generation diphenylbutylpiperidine antipsychotic.

Specifications

Chemical Name or Material Penfluridol
CAS 26864-56-2
Color White
Purity Grade Notes Research
Molecular Formula C28H27ClF5NO
Quantity 50 mg
Synonym R-16341
Solubility Information DMSO : ≥ 100 mg/mL (190.85 mM)
SMILES OC1(C2=CC=C(Cl)C(C(F)(F)F)=C2)CCN(CCCC(C3=CC=C(F)C=C3)C4=CC=C(F)C=C4)CC1
Molecular Weight (g/mol) 523.97
Formula Weight 523.97
Percent Purity 98.92%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.