Learn More
Pendimethalin, TRC
Specifications
Specifications
| Chemical Name or Material | Pendimethalin |
| CAS | 40487-42-1 |
| Molecular Formula | C13 H19 N3 O4 |
| Quantity | 1 g |
| Synonym | N-(1-Ethylpropyl)-3,4-dimethyl-2,6-dinitrobenzenamine, 3,4-Dimethyl-2,6-dinitro-N-(1-ethylpropyl)aniline, 3,4-Dimethyl-2,6-dinitro-N-(1'-ethylpropyl)aniline, AC 92553, Accotab, Activus EC, Acumen, Gogosan, Herbadox, Most Micro, N-(1-Ethylpropyl)-2,6-dinitro-3,4-xylidine, N-(1-Ethylpropyl)-3,4-dimethyl-2,6-dinitroaniline, OWC, Ornamental Weedgrass Control, Panida, Panida 330EC, Pendimax, Pendimethalin, Pendimethaline, Pendulum, Penoxalin, Penoxaline, Phenoxalin, Prowl, Prowl 3.3, Prowl 400, SWGC, Southern Weed Grass Control, Stomp, Stomp 330, Stomp 330D, Stomp 330E, Stomp 330EC, Benzenamine','N-(1-ethylpropyl)-3,4-dimethyl-2,6-dinitro- |
| InChI Formula | InChI=1S/C13H19N3O4/c1-5-10(6-2)14-12-11(15(17)18)7-8(3)9(4)13(12)16(19)20/h7,10,14H,5-6H2,1-4H3 |
| SMILES | CCC(CC)Nc1c(cc(C)c(C)c1[N+](=O)[O-])[N+](=O)[O-] |
| IUPAC Name | 3,4-dimethyl-2,6-dinitro-N-pentan-3-ylaniline |
| Molecular Weight (g/mol) | 281.31 |
| Formula Weight | 281.1376 |