missing translation for 'onlineSavingsMsg'
Learn More

Pemigatinib, MedChemExpress

Product Code. p-200100695
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16486997

Brand: Medchem Express HY109099

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pemigatinib (INCB054828) is an orally active, selective FGFR inhibitor with IC50s of 0.4 nM, 0.5 nM, 1.2 nM, 30 nM for FGFR1, FGFR2, FGFR3, FGFR4, respectively. Pemigatinib has the potential for cholangiocarcinoma.

Pemigatinib (INCB054828) is an orally active, selective FGFR inhibitor with IC50s of 0.4 nM, 0.5 nM, 1.2 nM, 30 nM for FGFR1, FGFR2, FGFR3, FGFR4, respectively. Pemigatinib has the potential for cholangiocarcinoma.

Specifications

Chemical Name or Material Pemigatinib
CAS 1513857-77-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H27F2N5O4
Quantity 5 mg
Synonym INCB054828
Solubility Information DMSO : 25 mg/mL (51.28 mM; ultrasonic and warming and heat to 60°C)
SMILES CCN(C(N(C(C(F)=C(OC)C=C1OC)=C1F)C2)=O)C(C2=CN3)=C4C3=NC(CN5CCOCC5)=C4
Molecular Weight (g/mol) 487.5
Formula Weight 487.5
Percent Purity 98.84%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.