Learn More
Pemigatinib, MedChemExpress
Product Code. 16486997
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16486997
Brand: Medchem Express HY109099
MedChemExpress Pemigatinib (INCB054828) is an orally active, selective FGFR inhibitor with IC50s of 0.4 nM, 0.5 nM, 1.2 nM, 30 nM for FGFR1, FGFR2, FGFR3, FGFR4, respectively. Pemigatinib has the potential for cholangiocarcinoma.
Pemigatinib (INCB054828) is an orally active, selective FGFR inhibitor with IC50s of 0.4 nM, 0.5 nM, 1.2 nM, 30 nM for FGFR1, FGFR2, FGFR3, FGFR4, respectively. Pemigatinib has the potential for cholangiocarcinoma.
Specifications
Chemical Name or Material Pemigatinib
CAS 1513857-77-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H27F2N5O4
Quantity 5 mg
Synonym INCB054828
Solubility Information DMSO : 25 mg/mL (51.28 mM; ultrasonic and warming and heat to 60°C)
SMILES CCN(C(N(C(C(F)=C(OC)C=C1OC)=C1F)C2)=O)C(C2=CN3)=C4C3=NC(CN5CCOCC5)=C4
Molecular Weight (g/mol) 487.5
Formula Weight 487.5
Percent Purity 98.84%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.