Learn More
3M™ PELTOR™ Optime™ II Helmet Mounted Ear Muffs
Click to view available options
Color:
Black
Unit Size:
Each
Pack of 20
Specifications
Specifications
| Color | Black |
| Certifications/Compliance | CE Approved |
| For Use With (Application) | Suitable Choice for Environments With Substantial Industrial Noise or Construction Machinery, Like Airports and Agricultural Work |
| Noise Reduction Rating (NRR) | 30 dB |
| Plug Shape | Oval |
| Product Type | Ear Muffs Optime II |
| Dielectric | Yes |
| Material | Stainless Steel |
| Style | Helmet-mounted |
| Includes | P3E Helmet Attachment for Helmet Mounting |