Learn More
3M™ PELTOR™ Optime™ II Earmuffs
Product Code. p-7160321
Change view
Click to view available options
Model:
H520A-472-GB
H520F-409-GQ
H520F-460-GB
H520P3A-410-GQ
H520P3AF-410-GQ
H520P3B-410-GQ
H520P3E-410-GQ-01
H520P3E-467-GB
H520P3EA-410-GQ
H520P3F-410-GQ
H520P3G-410-GQ
H520P3H-410-GQ
H520P3H-467-GB
H520P3K-410-GQ
Unit Size:
Pack of 10
Pack of 20
Pack of 6
Product Code. Model unitSize
15710028 H520P3G-410-GQ Pack of 20
15719697 H520A-472-GB Pack of 10
15719917 H520P3A-410-GQ Pack of 20
15729697 H520F-409-GQ Pack of 10
15729917 H520P3B-410-GQ Pack of 20
15739697 H520F-460-GB Pack of 10
15739857 H520P3H-467-GB Pack of 10
15739917 H520P3E-410-GQ-01 Pack of 20
15749697 H520P3E-467-GB Pack of 10
15749917 H520P3EA-410-GQ Pack of 20
15759917 H520P3F-410-GQ Pack of 20
15769917 H520P3H-410-GQ Pack of 20
15779917 H520P3K-410-GQ Pack of 20
15799727 H520P3AF-410-GQ Pack of 6
14 options
Product Code. 15710028
Brand: 3M™ 7100006656
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ideal for environments with substantial industrial noise or construction machinery, like airports and agricultural work. 3M™ PELTOR™ Optime™ II Earmuffs have been developed for demanding noise-hazard environments.
  • Ideal hearing protection against noise arising from environments with substantial industrial noise or construction machinery, like airports and agricultural work
  • Soft wide cushions helps reduce pressure around the ears and improves comfort and wearability
  • Large space inside cup helps reduce moisture and heat build-up
  • Easy to replace cushions and inserts helps keep them hygienically clean
TRUSTED_SUSTAINABILITY
Specifications
Color Green
Certifications/Compliance CE Approved, EN 352-3: 2002
Model H520P3G-410-GQ
Noise Reduction Rating (NRR) 30 dB
Product Type Ear Muffs
Dielectric No