Learn More
3M™ PELTOR™ Optime™ II Earmuffs
Description
- Ideal hearing protection against noise arising from environments with substantial industrial noise or construction machinery, like airports and agricultural work
- Soft wide cushions helps reduce pressure around the ears and improves comfort and wearability
- Large space inside cup helps reduce moisture and heat build-up
- Easy to replace cushions and inserts helps keep them hygienically clean
Specifications
Specifications
| Color | Green |
| Certifications/Compliance | CE Approved, EN 352-3: 2002 |
| Model | H520P3G-410-GQ |
| Noise Reduction Rating (NRR) | 30 dB |
| Product Type | Ear Muffs |
| Dielectric | No |