Learn More
3M™ PELTOR™ Optime™ I Earmuffs
Product Code. 15749907
Change view
Click to view available options
Model:
H510A-470-GB
H510B-403-GU
H510F-404-GU
H510F-459-GB
H510P3A-405- GU
H510P3A-405-GU
H510P3AF-405-GU
H510P3B-405-GU
H510P3E-405-GU
H510P3E-469-GB
H510P3EA-405-GU
H510P3G-405-GU
H510P3H-405-GU
H510P3K-405-GU
Style:
Hard Hat
Neckband
Over-the-head
Unit Size:
Each
Pack of 10
Pack of 20
Pack of 6
Product Code. Model Style unitSize
15749907 H510P3H-405-GU Hard Hat Pack of 20
15709697 H510P3E-469-GB Hard Hat Pack of 10
15709907 H510P3A-405- GU Hard Hat Pack of 20
15719707 H510P3E-405-GU Hard Hat Pack of 20
15719857 H510A-470-GB Over-the-head Each
15719907 H510P3B-405-GU Hard Hat Pack of 20
17281806 H510B-403-GU Neckband Pack of 10
15729857 H510F-459-GB Over-the-head Pack of 10
15729907 H510P3EA-405-GU Hard Hat Pack of 20
15739907 H510P3G-405-GU Hard Hat Pack of 20
15759907 H510P3K-405-GU Hard Hat Pack of 20
15769907 H510P3A-405-GU Hard Hat Pack of 20
15789727 H510P3AF-405-GU Hard Hat Pack of 6
15799687 H510F-404-GU Over-the-head Pack of 10
14 options
Product Code. 15749907
Brand: 3M™ 7000107751
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
PELTOR™ Optime™ I Ear Muffs are very lightweight, providing high wearer comfort. Ideal for protection against noise arising from a wide range of applications in the workplace and leisure activities. Offer versatile protection for both short and long duration tasks.
  • Low-profile design which minimizes obtrusiveness
  • Large space inside cup helps reduce moisture and heat build-up
  • Easy-to-replace cushions and inserts helps keep them hygienically clean
  • Soft, wide cushions help reduce pressure around the ears and improve comfort and wearability
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow
Certifications/Compliance CE Approved, EN 352-3: 2002
Model H510P3H-405-GU
Noise Reduction Rating (NRR) 26 dB
Product Type Ear Muffs
Style Hard Hat